Erbium triacetate tetrahydrate ≥99% (REO, rare earth oxide basis)
Supplier: Thermo Scientific Chemicals
Synonyms:
Erbium(III) acetate tetrahydrate
045897-14
045897-22
AA45897-14EA
79.48
USD
AA45897-14
AA45897-22
Erbium triacetate tetrahydrate ≥99% (REO, rare earth oxide basis)
Erbium triacetate tetrahydrate
Purity: 99%
CAS No.: 15280-57-6
Molecular Formula: C6H9ErO6.4H2O
Formula Weight: 416.45 (344.39anhy)
Physical Form: Crystalline Aggregates
Density: 2.114
Appearance: Pink
Odor: Odorless
MDL No.: MFCD00013034
Formula:
C₆H₉ErO₆·4H₂O MW: 416.45 g/mol Density: 2,114 Storage Temperature: Ambient |
MDL Number:
MFCD00013034 CAS Number: 15280-57-6 EINECS: 247-067-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...