Molybdenum ≥99% (metals basis), nanopowder APS 60-80 nm
Supplier: Thermo Scientific Chemicals
|
Danger
|
045794-06
045794-14
AA45794-06EA
200
USD
AA45794-06
AA45794-14
Molybdenum ≥99% (metals basis), nanopowder APS 60-80 nm
Molybdenum
Purity: 99%
CAS No.: 7439-98-7
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00003465
Formula:
Mo MW: 95.94 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003465 CAS Number: 7439-98-7 EINECS: 231-107-2 UN: 3089 ADR: 4.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...