Dimethyl acetylenedicarboxylate ≥95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dimethyl but-2-yne-1,4-dioate, Dimethyl but-2-ynedioate
045131-14
045131-22
045131-36
AA45131-14EA
79.12
USD
AA45131-14
AA45131-22
AA45131-36
Dimethyl acetylenedicarboxylate ≥95%
Dimethyl acetylenedicarboxylate
Purity: 95%
CAS No.: 762-42-5
Molecular Formula: C6H6O4
Formula Weight: 142.11
Flash Point: 86°(186°F)
Physical Form: Liquid
Density: 1.156
Refractive Index: 1.4470
Sensitivity: Light Sensitive
Appearance: Colorless to pale yellow
Melting Point: -18°
Boiling Point: 95-98°/19mm
Beilstein Reference: 607063
UN No.: 1760
MDL No.: MFCD00008456
Formula:
C₆H₆O₄ MW: 142.11 g/mol Boiling Pt: 95…98 °C (19 mmHg) Melting Pt: –18 °C Density: 1.156 Flash Pt: 86 °C (186 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008456 CAS Number: 762-42-5 EINECS: 212-098-4 UN: 1760 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...