Indium ≥99.9% (metals basis), Powder -100 mesh

Supplier: Thermo Scientific Chemicals
Warning

045122-06 045122-14
AA45122-06EA 106.72 USD
AA45122-06 AA45122-14
Indium ≥99.9% (metals basis), Powder -100 mesh
Indium

Purity: 99.9% CAS No.: 7440-74-6 Molecular Formula: In Formula Weight: 114.82 Physical Form: Powder Density: 7.31 Appearance: Silver-grey Odor: Odorless Melting Point: 156.6° Boiling Point: 2072° UN No.: 3089 MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2072 °C
Melting Pt: 156.6 °C
Density: 7.31 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
UN: 3089
ADR: 4.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR