Manganese ≥99.6% (metals basis), powder APS <10 µm

Supplier: Thermo Scientific Chemicals
Warning

045098-18 045098-30 045098-A1
AA45098-18EA 37.2 USD
AA45098-18 AA45098-30 AA45098-A1
Manganese ≥99.6% (metals basis), powder APS <10 µm
Manganese

Purity: 99.6% CAS No.: 7439-96-5 UN No.: 3089 MDL No.: MFCD00011111

Formula: Mn
Boiling Pt: 2097 °C (1013 hPa)
Melting Pt: 1260 °C
Density: 7.2 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011111
CAS Number: 7439-96-5
EINECS: 231-105-1
UN: 3089
ADR: 4.1,III
Merck Index: 12,05762

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR