Glassy carbon, rods, Diameter 6 mm (0.24 in) Type 2
Supplier: Thermo Scientific Chemicals
045005-DJ
045005-DM
AA45005-DJEA
404
USD
AA45005-DJ
AA45005-DM
AA45005-DR
Glassy carbon, rods, Diameter 6 mm (0.24 in) Type 2
Glassy carbon
Physical Form: ≈0.40g/cm
MDL No.: MFCD00133992
Formula:
C Melting Pt: 3652 °C Density: 1.95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00133992 CAS Number: 7440-44-0 EINECS: 231-153-3 Merck Index: 13,01818 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...