tetra-Chloroauric(III) acid (40 - 44% Au) (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Auric chloride, tetra-Chloroauric acid, Gold chloride, Gold trichloride, Hydrogen tetrachloroaurate (III)

044744-NB 044744-NH
AA44744-NBEA 1240 USD
AA44744-NB AA44744-NH
tetra-Chloroauric(III) acid (40 - 44% Au) (w/w) in aqueous solution
tetra-Chloroauric(III) acid

CAS No.: 16903-35-8 Molecular Formula: HAuCl4 Formula Weight: 339.79 Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Orange UN No.: 3264 MDL No.: MFCD00011322

Formula: HAuCl₄
MW: 339.78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011322
CAS Number: 16903-35-8
EINECS: 240-948-4
UN: 3264
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR