Bismuth(III) citrate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Bismuth citrate
040398-30
040398-A1
AA40398-30EA
109.6
USD
AA40398-30
AA40398-A1
Bismuth(III) citrate 94%
Bismuth(III) citrate
Insoluble in water, alcohol, ether. Soluble in ammonia solution and in solution of alkali citrates. Typical tap density 0.50g/cm?
Purity: 94%
CAS No.: 813-93-4
Molecular Formula: C6H5BiO7
Formula Weight: 398.08
Physical Form: Crystalline Powder, typically 10 micron max
Density: 3.5
Appearance: White
Odor: Odorless
Melting Point: dec.
Solubility: Insoluble in water, alcohol, ether. Soluble in ammonia solution and in solution of alkali citrates. Typical tap density 0.50g/cm3
MDL No.: MFCD00050817
Formula:
C₆H₅BiO₇ MW: 398.08 g/mol Melting Pt: Dec. Density: 3,5 Storage Temperature: Ambient |
MDL Number:
MFCD00050817 CAS Number: 813-93-4 EINECS: 212-390-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...