Tungsten(VI) ethoxide 5% (w/v) in ethanol , 99.8% (metals basis)

Supplier: Thermo Scientific Chemicals
Danger

039170-AE 039170-AC
AA39170-AEEA 336 USD
AA39170-AE AA39170-AC
Tungsten(VI) ethoxide 5% (w/v) in ethanol , 99.8% (metals basis)
Tungsten (VI) ethoxide

Purity: 99.8% CAS No.: 62571-53-3 Molecular Formula: C12H30O6W Formula Weight: 454.21 Flash Point: 17°(63°F) Physical Form: Liquid Sensitivity: Moisture Sensitive Appearance: Pale yellow to brown Odor: Alcohol-like UN No.: 1170 MDL No.: MFCD00156526

Formula: C₁₀H₂₅O₅W
MW: 454.21 g/mol
Flash Pt: 17 °C (63 °F)
Storage Temperature: Ambient
MDL Number: MFCD00156526
CAS Number: 62571-53-3
UN: 1170
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR