Selenourea ≥99.97% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
036720-03
036720-06
AA36720-03EA
107.52
USD
AA36720-03
AA36720-06
Selenourea ≥99.97% (metals basis)
Selenourea
Purity: 99.97%
CAS No.: 630-10-4
Molecular Formula: (H2N)2C=Se
Formula Weight: 123.02
Physical Form: Powder, Ampouled under argon
Sensitivity: Air and Moisture Sensitive
Appearance: White to grey
Melting Point: ca 210° dec.
UN No.: 3283
MDL No.: MFCD00008065
Formula:
CH₄N₂Se MW: 123.02 g/mol Melting Pt: ∼ 210 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008065 CAS Number: 630-10-4 EINECS: 211-129-9 UN: 3283 ADR: 6.1,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...