Tin(II) acetate ≥95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Decomposes in water. Soluble in dilute HCl
Purity: 95%
CAS No.: 638-39-1
Molecular Formula: Sn(OOCCH3)2
Formula Weight: 236.80
Physical Form: Powder/Lump
Density: 2.31
Appearance: White
Melting Point: 182-183°
Solubility: Decomposes in water. Soluble in dilute HCl
Merck Reference: 14,8781
UN No.: 3146
MDL No.: MFCD00054408
Formula:
C₄H₆O₄Sn Melting Pt: 182…183 °C Density: 2.31 Storage Temperature: Ambient |
MDL Number:
MFCD00054408 CAS Number: 638-39-1 EINECS: 211-335-9 UN: 3146 ADR: 6.1,III Merck Index: 14,8781 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...