Ammonium carbamate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Carbamic acid ammonium salt
018134-30
018134-18
AA18134-30EA
128
USD
AA18134-30
AA18134-18
Ammonium carbamate 98%
Ammonium carbamate
Ammoniating agent
Freely soluble in water. Soluble in alcohol
Purity: 98%
CAS No.: 1111-78-0
Molecular Formula: CH6N2O2
Formula Weight: 78.07
Physical Form: Powder
Density: 1.6
Appearance: Off-white
Melting Point: 60° subl.
Solubility: Freely soluble in water. Soluble in alcohol
Beilstein Reference: 3914091
Merck Reference: 14,507
MDL No.: MFCD00013010
Ammoniating agent
Formula:
CH₆N₂O₂ MW: 78.07 g/mol Melting Pt: 60 °C (subl.) Density: 1.6 Storage Temperature: Ambient |
MDL Number:
MFCD00013010 CAS Number: 1111-78-0 EINECS: 214-185-2 Merck Index: 14,507 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...