2-Ethoxy-3-pyridineboronic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (2-Ethoxypyridin-3-yl)boronic acid

L20422-06 L20422-03 L20422-MD
AAAL20422-06EA 693 USD
AAAL20422-06 AAAL20422-03 AAAL20422-MD
2-Ethoxy-3-pyridineboronic acid 98%
2-Ethoxy-3-pyridineboronic acid

Purity: 98% CAS No.: 854373-97-0 Molecular Formula: C7H10BNO3 Formula Weight: 166.97 MDL No.: MFCD06201036

Formula: C₇H₁₀BNO₃
MW: 166.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06201036
CAS Number: 854373-97-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR