Phenylselenocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

L20194-06 L20194-03
AAAL20194-06EA 120.77 USD
AAAL20194-06 AAAL20194-03
Phenylselenocyanate 98%
Phenylselenocyanate

Fieser: 7,253 8,119 9,34 11,33


Purity: 98% CAS No.: 2179-79-5 Molecular Formula: C7H5NSe Formula Weight: 182.08 Flash Point: 110°(230°F) Refractive Index: 1.6030 Boiling Point: 116-117°/12mm Beilstein Reference: 1617370 UN No.: 3440 MDL No.: MFCD00216944

Formula: C₇H₅NSe
Boiling Pt: 116…117 °C/12 mm
Density: 1.484
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00216944
CAS Number: 2179-79-5
UN: 3440
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR