2-Isopropylphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20110-03
L20110-06
AAAL20110-03EA
111
USD
AAAL20110-03
AAAL20110-MD
AAAL20110-06
2-Isopropylphenylboronic acid 97%
2-Isopropylphenylboronic acid
Purity: 97%
CAS No.: 89787-12-2
Molecular Formula: C9H13BO2
Formula Weight: 164.01
Melting Point: 85-86°
Beilstein Reference: 3090298
Formula:
C₉H₁₃BO₂ Melting Pt: 85…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411937 CAS Number: 89787-12-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...