6-(Trifluoromethyl)nicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L20054-MD L20054-03
AAAL20054-MDEA 69.2 USD
AAAL20054-MD AAAL20054-03
6-(Trifluoromethyl)nicotinic acid 97%
6-(Trifluoromethyl)nicotinic acid

Purity: 97% CAS No.: 231291-22-8 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Melting Point: 191-192° MDL No.: MFCD00792430

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 191…192 °C
Storage Temperature: Ambient
MDL Number: MFCD00792430
CAS Number: 231291-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR