N-Boc-ethylenediamine 98% may cont up to 5% tert-butanol
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(tert-Butoxycarbonyl)-1,2-diaminoethane, N-(tert-Butoxycarbonyl)-1,2-ethylenediamine, tert-Butyl (2-aminoethyl)carbamate, t-boc-N-EDA, tert-Butyl N-(2-Aminoethyl)carbamate, N-BOC-1,2-diaminoethane
L19947-06
L19947-14
AAAL19947-06EA
135.99
USD
AAAL19947-06
AAL19947-14
N-Boc-ethylenediamine 98% may cont up to 5% tert-butanol
N-Boc-ethylenediamine
Purity: 98%
CAS No.: 57260-73-8
Molecular Formula: (CH3)3COCONHCH2CH2NH2
Formula Weight: 160.22
Flash Point:
Formula:
C₇H₁₆N₂O₂ MW: 160.22 g/mol Boiling Pt: 72 °C/0.1 mm Density: 1.016 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00191871 CAS Number: 57260-73-8 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...