4-Cyano-2-fluorobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L19722-06 L19722-14
AAAL19722-03EA 38.63 USD
AAAL19722-03 AAAL19722-06 AAAL19722-14
4-Cyano-2-fluorobenzoic acid 98%
4-Cyano-2-fluorobenzoic acid

Purity: 98% CAS No.: 164149-28-4 Molecular Formula: C8H4FNO2 Formula Weight: 165.12 Melting Point: 214-218° Beilstein Reference: 8051929 MDL No.: MFCD03094454

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 214…218 °C
Storage Temperature: Ambient
MDL Number: MFCD03094454
CAS Number: 164149-28-4
UN: 3439
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR