(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

L19679-03 L19679-MD
AAAL19679-03EA 264 USD
AAAL19679-03 AAAL19679-MD
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine 97%
(R)-(+)-N,N-Dimethyl-1-ferrocenylethylamine

Purity: 97% CAS No.: 31886-58-5 Molecular Formula: C14H19FeN Formula Weight: 257.16 Flash Point: 110°(230°F) Refractive Index: 1.5890 Boiling Point: 120-121°/0.7mm Optical Rotation: +13° (c=1 in ethanol, 5H) MDL No.: MFCD00066273

Formula: C₁₄H₁₉FeN
MW: 257.16 g/mol
Boiling Pt: 120…121 °C/0.7 mm
Density: 1.222
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00066273
CAS Number: 31886-58-5

Order Now

Specification Test Results

Optical rotation +13° (c=1 in ethanol, 5H)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR