Isoquinoline-1-carbonitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Isoquinolinecarbonitrile
L19425-03
L19425-06
AAAL19425-03EA
65.3
USD
AAAL19425-03
AAAL19425-06
Isoquinoline-1-carbonitrile 99%
Isoquinoline-1-carbonitrile
Purity: 99%
CAS No.: 1198-30-7
Molecular Formula: C10H6N2
Formula Weight: 154.17
Odor: Odorless
Melting Point: 87-91°
Beilstein Reference: 119104
MDL No.: MFCD00134166
Formula:
C₁₀H₆N₂ MW: 154.17 g/mol Melting Pt: 87…91 °C Storage Temperature: Ambient |
MDL Number:
MFCD00134166 CAS Number: 1198-30-7 UN: 3439 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...