5-Bromo-2-methyl-2-pentene 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Bromo-2-methylpent-2-ene
L19406-06
L19406-03
AAAL19406-06EA
232
USD
AAAL19406-06
AAAL19406-03
5-Bromo-2-methyl-2-pentene 97%
5-Bromo-2-methyl-2-pentene
Purity: 97%
CAS No.: 2270-59-9
Molecular Formula: C6H11Br
Formula Weight: 163.06
Storage Temperature: 2°C to 8°C
Flash Point: 23°(73°F)
Refractive Index: 1.4775
Boiling Point: 50°/20mm
Beilstein Reference: 1735749
UN No.: 1993
MDL No.: MFCD00009887
Formula:
C₆H₁₁Br MW: 163.06 g/mol Boiling Pt: 50 °C/20 mm Density: 1.219 Flash Pt: 23 °C(73 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009887 CAS Number: 2270-59-9 UN: 1993 ADR: "3",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...