1-Benzylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

L19339-18 L19339-09
AAAL19339-18EA 335 USD
AAAL19339-18 AAAL19339-09
1-Benzylpiperidine 98%
1-Benzylpiperidine

Purity: 98% CAS No.: 2905-56-8 Molecular Formula: C12H17N Formula Weight: 175.28 Refractive Index: 1.5260 Boiling Point: 120-123°/9mm Beilstein Reference: 5639 MDL No.: MFCD00224901

Formula: C₁₂H₁₇N
MW: 175.28 g/mol
Boiling Pt: 120…123 °C/9 mm
Density: 0.950
Storage Temperature: Ambient
MDL Number: MFCD00224901
CAS Number: 2905-56-8
EINECS: 220-809-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR