(R)-(-)-2-Aminohexane ≥99%, ee 96%, ChiPros®

Supplier: Thermo Scientific Chemicals
Danger

L19304-14 L19304-06
AAAL19304-14EA 652 USD
AAAL19304-14 AAAL19304-06
(R)-(-)-2-Aminohexane ≥99%, ee 96%, ChiPros®
(R)-(-)-2-Aminohexane

Purity: 99+% CAS No.: 70095-40-8 Molecular Formula: C6H15N Formula Weight: 101.19 Flash Point: -3°(26°F) Melting Point: -20° Boiling Point: 116° Beilstein Reference: 1718994 UN No.: 2733 MDL No.: MFCD00671627

Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 116 °C
Melting Pt: < –20 °C
Density: 0.755
Flash Pt: -3 °C(26 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671627
CAS Number: 70095-40-8
EINECS: 622-790-1
UN: 2733
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR