6-Methoxy-2-naphthaleneboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Methoxy-2-naphthylboronic acid, 6-Methoxy-2-naphthylboronic Acid
L19060-03
L19060-06
AAAL19060-03EA
31.1
USD
AAAL19060-03
AAAL19060-06
6-Methoxy-2-naphthaleneboronic acid 95%
6-Methoxy-2-naphthaleneboronic acid
Purity: 95%
CAS No.: 156641-98-4
Molecular Formula: C11H11BO3
Formula Weight: 202.02
Melting Point: ca 245° dec.
Beilstein Reference: 7636998
Formula:
C₁₁H₁₁BO₃ MW: 202.02 g/mol Melting Pt: ca 245 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03093087 CAS Number: 156641-98-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...