1-(2-Tetrahydrofuroyl)piperazine 97%

Supplier: Thermo Scientific Chemicals
Danger

L18704-06 L18704-14
AAAL18704-06EA 43.5 USD
AAAL18704-06 AAAL18704-14
1-(2-Tetrahydrofuroyl)piperazine 97%
1-(2-Tetrahydrofuroyl)piperazine

Purity: 97% CAS No.: 63074-07-7 Molecular Formula: C9H16N2O2 Formula Weight: 184.24 Boiling Point: 120-125°/0.2mm Beilstein Reference: 880578

Formula: C₉H₁₆N₂O₂
Boiling Pt: 120…125 °C/0.2 mm
Storage Temperature: Ambient
MDL Number: MFCD00190247
CAS Number: 63074-07-7
EINECS: 613-137-1
UN: 3267
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR