2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite 95%

Supplier: Thermo Scientific Chemicals
Danger

L18575-03 L18575-06
AAAL18575-03EA 125 USD
AAAL18575-03 AAAL18575-06
2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite 95%
2-Cyanoethyl-N,N-diisopropylchlorophosphoramidite

Purity: 95% CAS No.: 89992-70-1 Molecular Formula: C9H18ClN2OP Formula Weight: 236.68 Storage Temperature: -30°C to -10°C Flash Point: 110°(230°F) Refractive Index: 1.4780 Boiling Point: 103-105°/0.08mm Beilstein Reference: 3588525 UN No.: 2845 MDL No.: MFCD00011544

Formula: C₉H₁₈ClN₂OP
MW: 236.68 g/mol
Boiling Pt: 103…105 °C/0.08 mm
Density: 1.061
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00011544
CAS Number: 89992-70-1
UN: 2845
ADR: "4.2",I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR