4-sulfamidobenzoyl chloride DMF complex 95%

Supplier: Thermo Scientific Chemicals
Danger

L18428-06 L18428-14
AAAL18428-06EA 103 USD
AAAL18428-06 AAL18428-14
4-sulfamidobenzoyl chloride DMF complex 95%
4-sulfamidobenzoyl chloride DMF complex

Purity: 95% CAS No.: 1172493-99-0 Molecular Formula: C10H13ClN2O4S Formula Weight: 292.74 UN No.: 3261 MDL No.: MFCD02094019

Formula: C₁₀H₁₃ClN₂O₄S
MW: 292.74 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02094019
CAS Number: 1172493-99-0
UN: 3261
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR