4-Methyl-3-nitrobenzamide 98%

Supplier: Thermo Scientific Chemicals
Warning

L18270-06 L18270-14
AAAL18270-06EA 17.6 USD
AAAL18270-06 AAAL18270-14
4-Methyl-3-nitrobenzamide 98%
4-Methyl-3-nitrobenzamide

Purity: 98% CAS No.: 19013-11-7 Molecular Formula: C8H8N2O3 Formula Weight: 180.16 Melting Point: 164-167° Beilstein Reference: 2102644

Formula: C₈H₈N₂O₃
MW: 180.16 g/mol
Melting Pt: 164…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00027392
CAS Number: 19013-11-7
EINECS: 242-752-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR