Ethyl-4-acetylbutyrate 97%
Supplier: Thermo Scientific Chemicals
Warning
|
Synonyms:
Ethyl 4-Acetylbutyrate
L17848-14
L17848-22
L17848-06
AAAL17848-14EA
140
USD
AAAL17848-14
AAAL17848-22
AAAL17848-06
Ethyl-4-acetylbutyrate 97%
Ethyl-4-acetylbutyrate
Purity: 97%
CAS No.: 13984-57-1
Molecular Formula: C8H14O3
Formula Weight: 158.20
Flash Point: 69°(156°F)
Refractive Index: 1.4275
Boiling Point: 65°/0.76mm
Beilstein Reference: 1762625
MDL No.: MFCD00009213
Formula:
C₈H₁₄O₃ Boiling Pt: 65 °C/0.76 mm Density: 0.989 Flash Pt: 69 °C(156 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00009213 CAS Number: 13984-57-1 EINECS: 237-776-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...