Fomblin® YR-1800

Supplier: Thermo Scientific Chemicals

Synonyms: Perfluoropolyether YR-1800

L17548-06 L17548-14
AAAL17548-06EA 36.4 USD
AAAL17548-06 AAAL17548-14
Fomblin® YR-1800
1-Propene, 1,1,2,3,3,3-hexafluoro-, oxidized, polymerized

CAS No.: 69991-67-9 Refractive Index: 1.3040 MDL No.: MFCD01323324

Density: 1.920
Storage Temperature: Ambient
MDL Number: MFCD01323324
CAS Number: 69991-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR