Isoflurane 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether

L17315-14 L17315-06
AAAL17315-14EA 232.9 USD
AAAL17315-14 AAAL17315-06
Isoflurane 97%
Isoflurane

Purity: 97% CAS No.: 26675-46-7 Molecular Formula: CF3CHClOCHF2 Formula Weight: 184.49 Density: 1.496 Refractive Index: 1.3 Odor: Ether-like Boiling Point: 48-49° Beilstein Reference: 1852087 Merck Reference: 14,5175 MDL No.: MFCD00066609

Formula: C₃H₂ClF₅O
MW: 184.49 g/mol
Boiling Pt: 48…49 °C
Density: 1.496
Storage Temperature: Ambient
MDL Number: MFCD00066609
CAS Number: 26675-46-7
EINECS: 247-897-7
Merck Index: 14,5175

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR