3-(tert-Butyldimethylsiloxy)thiophenol 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17238-03
L17238-06
AAAL17238-03EA
71.8
USD
AAAL17238-03
AAAL17238-06
3-(tert-Butyldimethylsiloxy)thiophenol 95%
3-(tert-Butyldimethylsiloxy)thiophenol
Purity: 95%
CAS No.: 216393-56-5
Molecular Formula: C12H20OSSi
Formula Weight: 240.44
Refractive Index: 1.5210
Formula:
C₁₂H₂₀OSSi MW: 240.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD01318115 CAS Number: 216393-56-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...