(R)-(+)-2-Benzyloxypropionic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-2-(Benzyloxy)propanoic acid

L17205-06 L17205-22 L17205-14
AAAL17205-06EA 52.3 USD
AAAL17205-06 AAAL17205-22 AAAL17205-14
(R)-(+)-2-Benzyloxypropionic acid 98%
(R)-(+)-2-Benzyloxypropionic acid

Purity: 98% CAS No.: 100836-85-9 Molecular Formula: C10H12O3 Formula Weight: 180.20 Melting Point: 51-53° Beilstein Reference: 4675524

Formula: C₁₀H₁₂O₃
MW: 180.20 g/mol
Melting Pt: 51…53 °C
Storage Temperature: Ambient
MDL Number: MFCD06799065
CAS Number: 100836-85-9

Order Now

Specification Test Results

Optical rotation +90° (c=1 in IMS)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR