5-Methyl-1H-indole-2-carboxylic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L17145-03 L17145-06
AAAL17145-03EA 90.3 USD
AAAL17145-03 AAAL17145-06
5-Methyl-1H-indole-2-carboxylic acid 99%
5-Methyl-1H-indole-2-carboxylic acid

Purity: 99% CAS No.: 10241-97-1 Molecular Formula: C10H9NO2 Formula Weight: 175.19 Melting Point: ca 235° dec. Beilstein Reference: 144055 MDL No.: MFCD00047166

Formula: C10H9NO2
MW: 175.19 g/mol
Melting Pt: ca 235 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00047166
CAS Number: 10241-97-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR