α-Methyl-4-trifluoromethylbenzyl alcohol 96%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-[4-(Trifluoromethyl)phenyl]ethanol
L16878-03
L16878-06
AAAL16878-03EA
49.9
USD
AAAL16878-03
AAAL16878-06
α-Methyl-4-trifluoromethylbenzyl alcohol 96%
alpha-Methyl-4-trifluoromethylbenzyl alcohol
Purity: 97%
CAS No.: 1737-26-4
Molecular Formula: C9H9F3O
Formula Weight: 190.17
Flash Point: 98°(208°F)
Refractive Index: 1.4580
Boiling Point: 62-63°/0.5mm
Beilstein Reference: 2257808
MDL No.: MFCD00039618
Formula:
C₉H₉F₃O MW: 190.17 g/mol Boiling Pt: 62…63 °C/0.5 mm Density: 1.237 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039618 CAS Number: 1737-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...