(Heptafluorobutyro)phenone 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16777-14
L16777-06
AAAL16777-14EA
293
USD
AAAL16777-14
AAAL16777-06
AAAL16777-22
(Heptafluorobutyro)phenone 99%
(Heptafluorobutyro)phenone
Purity: 99%
CAS No.: 559-91-1
Molecular Formula: C10H5F7O
Formula Weight: 274.14
Refractive Index: 1.4140
Boiling Point: 170-172°
Beilstein Reference: 2056234
MDL No.: MFCD00077552
Formula:
C₁₀H₅F₇O Boiling Pt: 170…172 °C Density: 1.473 Storage Temperature: Ambient |
MDL Number:
MFCD00077552 CAS Number: 559-91-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...