4'-Chloro-2,2':6',2''-terpyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

L14728-03 L14728-MD
AAAL14728-03EA 152 USD
AAAL14728-03 AAAL14728-MD
4'-Chloro-2,2':6',2''-terpyridine 98%
4'-Chloro-2,2':6',2''-terpyridine

Purity: 98% CAS No.: 128143-89-5 Molecular Formula: C15H10ClN3 Formula Weight: 267.72 Melting Point: 148-152° Beilstein Reference: 3613386 MDL No.: MFCD00191930

Formula: C₁₅H₁₀ClN₃
MW: 267.716 g/mol
Melting Pt: 148…152 °C
Storage Temperature: Ambient
MDL Number: MFCD00191930
CAS Number: 128143-89-5
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR