Benzyl-(S)-(-)-lactate 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
Benzyl L-lactate, (S)-(-)-Lactic Acid Benzyl Ester, Benzyl (S)-Lactate
L14645-06
L14645-14
AAAL14645-06EA
59.6
USD
AAAL14645-06
AAAL14645-14
Benzyl-(S)-(-)-lactate 97%
Benzyl-(S)-(-)-lactate
Purity: 97%
CAS No.: 56777-24-3
Molecular Formula: C10H12O3
Formula Weight: 180.20
Flash Point:
Formula:
C₁₀H₁₂O₃ MW: 180.20 g/mol Boiling Pt: 105…109 °C/0.1 mm Density: 1.120 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00209546 CAS Number: 56777-24-3 |
Specification Test Results
Optical rotation | -12.9° (c=3.1 in methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...