Methyl-2-phenoxybenzoate 99%
Supplier: Thermo Scientific Chemicals
Synonyms:
Methyl 2-phenoxybenzoate
L14584-09
L14584-18
AAAL14584-09EA
31.1
USD
AAAL14584-09
AAAL14584-18
AAAL14584-30
Methyl-2-phenoxybenzoate 99%
Methyl 2-phenoxybenzoate
Purity: 99%
CAS No.: 21905-56-6
Molecular Formula: C14H12O3
Formula Weight: 228.25
Refractive Index: 1.5760
Boiling Point: 130°/0.5mm
MDL No.: MFCD03093064
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol Boiling Pt: 130 °C/0.5 mm Storage Temperature: Ambient |
MDL Number:
MFCD03093064 CAS Number: 21905-56-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...