Luteolin tetramethylether 97%

Supplier: Thermo Scientific Chemicals

Synonyms: 5,7,3',4'-Tetramethoxyflavone, 2-(3,4-Dimethoxyphenyl)-5,7-dimethoxy-4H-chromen-4-one

L14496-ME L14496-MC
AAAL14496-MEEA 257 USD
AAAL14496-ME AAAL14496-MC
Luteolin tetramethylether 97%
Luteolin tetramethylether

Purity: 97% CAS No.: 855-97-0 Molecular Formula: C19H18O6 Formula Weight: 342.35 Melting Point: 193-197° Beilstein Reference: 336320 MDL No.: MFCD00017558

Formula: C₁₉H₁₈O₆
Melting Pt: 193…197 °C
Storage Temperature: Ambient
MDL Number: MFCD00017558
CAS Number: 855-97-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR