S-Trityl-L-cysteine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
H-Cys(Trt)-OH
L14384-06
L14384-03
AAAL14384-06EA
62.92
USD
AAAL14384-06
AAAL14384-03
S-Trityl-L-cysteine 97%
S-Trityl-L-cysteine
Purity: 97%
CAS No.: 2799-07-7
Molecular Formula: C22H21NO2S
Formula Weight: 363.48
Melting Point: ca 185° dec.
Beilstein Reference: 2339626
Optical Rotation: +84° (c=1 in acetic acid)
MDL No.: MFCD00002611
Formula:
C₂₂H₂₁NO₂S MW: 363.5 g/mol Melting Pt: ca 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00002611 CAS Number: 2799-07-7 |
Specification Test Results
Optical rotation | +84° (c=1 in acetic acid) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...