Diethoxymethylvinylsilane 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diethoxy(methyl)(vinyl)silane

L14224-22 L14224-14
AAAL14224-22EA 138 USD
AAAL14224-22 AAAL14224-14 AAAL14224-06
Diethoxymethylvinylsilane 97%
Diethoxymethylvinylsilane

Purity: 97% CAS No.: 5507-44-8 Molecular Formula: C7H16O2Si Formula Weight: 160.29 Flash Point: 17°(62°F) Refractive Index: 1.400 Boiling Point: 133-134° Beilstein Reference: 970124 UN No.: 1993 MDL No.: MFCD00009067

Formula: C₇H₁₆O₂Si
MW: 160.29 g/mol
Boiling Pt: 133…134 °C
Density: 0.858
Flash Pt: 17 °C (62 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009067
CAS Number: 5507-44-8
EINECS: 226-850-4
UN: 1993
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR