Genkwanin, 7-methoxyapigenin 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14169-MB
L14169-MC
AAAL14169-MBEA
60.8
USD
AAAL14169-MB
AAAL14169-MC
Genkwanin, 7-methoxyapigenin 97%
Genkwanin, 7-methoxyapigenin
Purity: 97%
CAS No.: 437-64-9
Molecular Formula: C16H12O5
Formula Weight: 284.27
Melting Point: 288-290°
Beilstein Reference: 292549
MDL No.: MFCD00017452
Formula:
C₁₆H₁₂O₅ Melting Pt: 288…290 °C Storage Temperature: Ambient |
MDL Number:
MFCD00017452 CAS Number: 437-64-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...