3-(3-Pyridyl)propionic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14007-06
L14007-14
AAAL14007-06EA
73.1
USD
AAAL14007-06
AAAL14007-14
3-(3-Pyridyl)propionic acid 97%
3-(3-Pyridyl)propionic acid
Purity: 97%
CAS No.: 3724-19-4
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 156-159°
Beilstein Reference: 122232
MDL No.: MFCD00274197
Formula:
C₈H₉NO₂ Melting Pt: 156…159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00274197 CAS Number: 3724-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...