Perfluorotetradecanoic acid 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L13796-03
L13796-06
AAAL13796-03EA
30.3
USD
AAAL13796-03
AAAL13796-06
Perfluorotetradecanoic acid 96%
Perfluorotetradecanoic acid
Purity: 96%
CAS No.: 376-06-7
Molecular Formula: CF3(CF2)12CO2H
Formula Weight: 714.12
Melting Point: 130-132°
Boiling Point: 192°/60mm
UN No.: 3261
MDL No.: MFCD00153263
Formula:
C₁₄HF₂₇O₂ Boiling Pt: 192 °C/60 mm Melting Pt: 130…132 °C Storage Temperature: Ambient |
MDL Number:
MFCD00153263 CAS Number: 376-06-7 EINECS: 206-803-4 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...