3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Phthalimidopropionic acid, 3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)propanoic acid

L13535-03 L13535-06
AAAL13535-03EA 34.7 USD
AAAL13535-03 AAAL13535-06
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid 98%
3-(1,3-Dioxoisoindolin-2-yl)propanoic acid

Purity: 98% CAS No.: 3339-73-9 Molecular Formula: C11H9NO4 Formula Weight: 219.20 Melting Point: 151-153° Beilstein Reference: 15570 MDL No.: MFCD00023096

Formula: C₁₁H₉NO₄
Melting Pt: 151…153 °C
Storage Temperature: Ambient
MDL Number: MFCD00023096
CAS Number: 3339-73-9
EINECS: 222-083-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR