4-Chloro-2-fluorobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Chloro-2-fluorophenyl)methanamine hydrochloride

L13488-06 L13488-03
AAAL13488-06EA 81.5 USD
AAAL13488-06 AAAL13488-03
4-Chloro-2-fluorobenzylamine hydrochloride 97%
4-Chloro-2-fluorobenzylamine hydrochloride

Purity: 97% CAS No.: 202982-63-6 Molecular Formula: C7H7ClFN.HCl Formula Weight: 196.05 Melting Point: 243-245° MDL No.: MFCD00143286

Formula: C₇H₇ClFN·HCl
MW: 196.05 g/mol
Melting Pt: 243…245 °C
Storage Temperature: Ambient
MDL Number: MFCD00143286
CAS Number: 202982-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR