4-Hydroxy-2-(trifluoromethyl)pyrimidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(Trifluoromethyl)pyrimidin-4-ol
L13118-03
L13118-06
AAAL13118-03EA
63.9
USD
AAAL13118-03
AAAL13118-06
4-Hydroxy-2-(trifluoromethyl)pyrimidine 97%
4-Hydroxy-2-(trifluoromethyl)pyrimidine
Purity: 97%
CAS No.: 1546-80-1
Molecular Formula: C5H3F3N2O
Formula Weight: 164.09
Melting Point: 168-171°
Beilstein Reference: 778887
MDL No.: MFCD00151833
Formula:
C₅H₃F₃N₂O MW: 164.09 g/mol Melting Pt: 168…171 °C Storage Temperature: Ambient |
MDL Number:
MFCD00151833 CAS Number: 1546-80-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...