Triethylvinylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Vinyltriethylsilane
L12461-06
L12461-03
AAAL12461-06EA
108
USD
AAAL12461-06
AAAL12461-03
Triethylvinylsilane 97%
Triethylvinylsilane
Purity: 97%
CAS No.: 1112-54-5
Molecular Formula: C8H18Si
Formula Weight: 142.32
Flash Point: 25°(77°F)
Refractive Index: 1.4340
Boiling Point: 145-146°
Beilstein Reference: 1743830
UN No.: 1993
MDL No.: MFCD00041345
Formula:
C₈H₁₈Si MW: 142.32 g/mol Boiling Pt: 145…146 °C Density: 0.771 Flash Pt: 25 °C (77 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00041345 CAS Number: 1112-54-5 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...