Ethyl-2-nitropropionate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L12261-03
L12261-06
AAAL12261-03EA
29.4
USD
AAAL12261-03
AAAL12261-06
Ethyl-2-nitropropionate 96%
Ethyl-2-nitropropionate
Purity: 96%
CAS No.: 2531-80-8
Molecular Formula: C5H9NO4
Formula Weight: 147.13
Flash Point: 89°(192°F)
Refractive Index: 1.4210
Boiling Point: 190-195°
Beilstein Reference: 774734
MDL No.: MFCD00024818
Formula:
C₅H₉NO₄ Boiling Pt: 190…195 °C Density: 1.130 Flash Pt: 89 °C(192 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00024818 CAS Number: 2531-80-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...